Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-1H-Triazole, 99.5%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass Bottle
Plastic bottle
288-88-0
C2H3N3
69.06
MFCD00005228
NSPMIYGKQJPBQR-UHFFFAOYSA-N
1,2,4-triazole,4h-1,2,4-triazole,pyrrodiazole,s-triazole,1,2,4-1h-triazole,1h-1,2,4-triazol,unii-10ms0y1rdi,1,2,4 triazole,1,2,4-triazol,4h-1,2,4-triazole van
9257
CHEBI:35550
1H-1,2,4-triazole
C1=NC=NN1
288-88-0
C2H3N3
69.06
MFCD00005228
NSPMIYGKQJPBQR-UHFFFAOYSA-N
1,2,4-triazole,4h-1,2,4-triazole,pyrrodiazole,s-triazole,1,2,4-1h-triazole,1h-1,2,4-triazol,unii-10ms0y1rdi,1,2,4 triazole,1,2,4-triazol,4h-1,2,4-triazole van
9257
CHEBI:35550
1H-1,2,4-triazole
C1=NC=NN1
Water 0.2% max. (K.F.)
Infrared Spectrum Authentic
Packaging Glass Bottle
Quantity 5g
Flash Point 140°C
Boiling Point 260°C
Color White
Melting Point 119°C to 122°C
CAS Min % 99.4
CAS Max % 100.0
Assay Percent Range 99.4% min. (HPLC)
Beilstein 26, 13
Fieser 01,1188; 02,423
Merck Index 15, 9766
Solubility Information Solubility in water: 1250g/L (20°C). Other solubilities: soluble in chloroform,xylene,ethylacetate and,alcohol
Formula Weight 69.06
Physical Form Crystalline Powder and Flakes
Percent Purity 99.50%
Chemical Name or Material 1, 2, 4-1H-Triazole
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Suspected of damaging the unborn child.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pr

GHS Signal Word: Warning

EINECSNumber : 206-022-9