Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4,5-Tetrachlorobenzene, 98%

Catalog No. ac157780050
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 95-94-3 | C6H2Cl4 | 215.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-94-3
Molecular Formula C6H2Cl4
Molecular Weight (g/mol) 215.89
MDL Number MFCD00000549
InChI Key JHBKHLUZVFWLAG-UHFFFAOYSA-N
Synonym s-tetrachlorobenzene, benzene, 1,2,4,5-tetrachloro, benzene tetrachloride, rcra waste number u207, 1,2,4,5-tetrachlorbenzol, unii-5n27529kgh, ccris 766, benzene, tetrachloro, rcra waste no. u207, 1,2,4,5-tetrachlorbenzol russian
PubChem CID 7270
ChEBI CHEBI:36697
IUPAC Name 1,2,4,5-tetrachlorobenzene
SMILES C1=C(C(=CC(=C1Cl)Cl)Cl)Cl

Specifications

Melting Point 139°C to 142°C
Color Gray to Pink
CAS Min % 97.5
CAS Max % 100.0
Boiling Point 240°C to 246°C
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Solid
Assay Percent Range 98%
Linear Formula C6H2Cl4
Beilstein 05, 205
Formula Weight 215.89
Percent Purity 98%
Chemical Name or Material 1,2,4,5-Tetrachlorobenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-466-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.