Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2,4-Dichlorophenyl)cyclopropanecarboxylic Acid 95%, Thermo Scientific™

Catalog No. AC134920050
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84604-70-6
Molecular Formula C10H8Cl2O2
Molecular Weight (g/mol) 231.072
MDL Number MFCD00044817
InChI Key MIBWNJWOBDYPRS-UHFFFAOYSA-N
Synonym 1-2,4-dichlorophenyl cyclopropanecarboxylic acid, 1-2,4-dichlorophenyl cyclopropane-1-carboxylic acid, cyclopropanecarboxylicacid, 1-2,4-dichlorophenyl, dichlorophenylcyclopropanecarboxylicacid, 1-2,4-dichlorophenyl cyclopropanecarboxylicacid, 1-2,4-dichloro-phenyl-cyclopropanecarboxylic acid, 1-2,4-dichlorophenyl-cyclopropanecarboxylic acid, 1-2,4-dichlorophenyl-1-cyclopropanecarboxylic acid, 1-2,4-dichlorophenyl-cyclopropane carboxylic acid
PubChem CID 736103
IUPAC Name 1-(2,4-dichlorophenyl)cyclopropane-1-carboxylic acid
SMILES C1CC1(C2=C(C=C(C=C2)Cl)Cl)C(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 140.0°C to 147.0°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Quantity 5 g
Formula Weight 231.08
Percent Purity 95%
Physical Form Granular Crystalline Powder
Chemical Name or Material 1-(2, 4-Dichlorophenyl)cyclopropanecarboxylic acid, 95%
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.