Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-Triacetoxybenzene 95.0+%, TCI America™
SDP

Catalog No. T0976500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0976500G 500 g
T097625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0976500G Supplier TCI America Supplier No. T0976500G

Chemical Identifiers

CAS 613-03-6
Molecular Formula C12H12O6
Molecular Weight (g/mol) 252.222
MDL Number MFCD00008700
InChI Key AESFGSJWSUZRGW-UHFFFAOYSA-N
Synonym 1,2,4-triacetoxybenzene, benzene-1,2,4-triyl triacetate, hydroxyhydroquinone triacetate, 1,2,4-benzenetriol, triacetate, pyrogallol a, 2-hydroxyhydroquinone triacetate, 1,2,4-benzenetriacetate, unii-903t7h7cp1, triacetate d'hydroxyhydroquinone, 2,4-bis acetyloxy phenyl acetate
PubChem CID 69169
IUPAC Name (3,4-diacetyloxyphenyl) acetate
SMILES CC(=O)OC1=CC(=C(C=C1)OC(=O)C)OC(=O)C

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 500 g
Formula Weight 252.22
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4-Triacetoxybenzene
RTECSNumber DC4800000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.