Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,2,4-Tribromobenzene, 95%

Click to view available options
Quantity:
5g
25g
615-54-3
C6H3Br3
314.802
MFCD00000062
FWAJPSIPOULHHH-UHFFFAOYSA-N
benzene, 1,2,4-tribromo,unii-eoe1kfc260,benzene,1,2,4-tribromo,eoe1kfc260,dsstox_cid_4346,dsstox_rid_77377,dsstox_gsid_24346,1,2,4-tribomobenzene,benzene,2,4-tribromo,acmc-1bdp2
12002
1,2,4-tribromobenzene
C1=CC(=C(C=C1Br)Br)Br
615-54-3
C6H3Br3
314.802
MFCD00000062
FWAJPSIPOULHHH-UHFFFAOYSA-N
benzene, 1,2,4-tribromo,unii-eoe1kfc260,benzene,1,2,4-tribromo,eoe1kfc260,dsstox_cid_4346,dsstox_rid_77377,dsstox_gsid_24346,1,2,4-tribomobenzene,benzene,2,4-tribromo,acmc-1bdp2
12002
1,2,4-tribromobenzene
C1=CC(=C(C=C1Br)Br)Br
Applications
1,2,4-tribromobenzene is used in the synthesis of hyper branched poly(p-phenylene ethynylenes). It is used as cross-linking reagent during Pd-catalyzed cross-coupling of 2,5-diiodo-4-[(2-ethylhexyl)oxy]methoxybenzene and 1,4-diethynyl-2,5-bis-(octyloxy)benzene. It is a compound commonly used in organic synthesis reactions.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Flash Point >110°C (230°F)
Boiling Point 274°C to 276°C
Melting Point 41°C to 43°C
Quantity 25g
Beilstein 1934823
Formula Weight 314.8
Percent Purity 95%
Chemical Name or Material 1,2,4-Tribromobenzene

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : DC1956500

TSCA : TSCA