Learn More
1,2,4-Tribromobenzene, 95%, Thermo Scientific Chemicals
Description
1,2,4-tribromobenzene is used in the synthesis of hyper branched poly(p-phenylene ethynylenes). It is used as cross-linking reagent during Pd-catalyzed cross-coupling of 2,5-diiodo-4-[(2-ethylhexyl)oxy]methoxybenzene and 1,4-diethynyl-2,5-bis-(octyloxy)benzene. It is a compound commonly used in organic synthesis reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,2,4-tribromobenzene is used in the synthesis of hyper branched poly(p-phenylene ethynylenes). It is used as cross-linking reagent during Pd-catalyzed cross-coupling of 2,5-diiodo-4-[(2-ethylhexyl)oxy]methoxybenzene and 1,4-diethynyl-2,5-bis-(octyloxy)benzene. It is a compound commonly used in organic synthesis reactions.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 41°C to 43°C |
Boiling Point | 274°C to 276°C |
Flash Point | >110°C (230°F) |
Quantity | 5 g |
Beilstein | 1934823 |
Synonym | benzene, 1,2,4-tribromo, unii-eoe1kfc260, benzene,1,2,4-tribromo, eoe1kfc260, dsstox_cid_4346, dsstox_rid_77377, dsstox_gsid_24346, 1,2,4-tribomobenzene, benzene,2,4-tribromo, acmc-1bdp2 |
Solubility Information | Insoluble in water. |
Formula Weight | 314.8 |
Percent Purity | 95% |
Chemical Name or Material | 1,2,4-Tribromobenzene |
Safety and Handling
Hazard Category | H315-H319-H335-H500 |
Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 210-433-9 |
RTECSNumber | DC1956500 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.