Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1,2,4-Tribromobenzene 95.0+%, TCI America™

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Melting Point | 42°C |
Color | White-Yellow |
Quantity | 25 g |
Synonym | benzene, 1,2,4-tribromo, unii-eoe1kfc260, benzene,1,2,4-tribromo, eoe1kfc260, dsstox_cid_4346, dsstox_rid_77377, dsstox_gsid_24346, 1,2,4-tribomobenzene, benzene,2,4-tribromo, acmc-1bdp2 |
Formula Weight | 314.80 |
Percent Purity | ≥95.0% (GC) |
Physical Form | Crystalline Lumps |
Chemical Name or Material | 1,2,4-Tribromobenzene |
Safety and Handling
RTECSNumber | DC1956500 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction