Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinic acid, disodium salt hexahydrate, 97%

Catalog No. AC158750010
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg

CAS: 6106-21-4 | C4H16Na2O10 | 270.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6106-21-4
Molecular Formula C4H16Na2O10
Molecular Weight (g/mol) 270.142
InChI Key ZBTUYCUNQBRXOR-UHFFFAOYSA-L
Synonym disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate
PubChem CID 3083938
ChEBI CHEBI:63686
IUPAC Name disodium;butanedioate;hexahydrate
SMILES C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+]

Specifications

Melting Point 120.0°C
Color Brown to Yellow
pH 7 to 9 (0.4M soln.)
Loss on Drying 37.0 % to 41.0 % (90°C)
Infrared Spectrum Authentic
Assay Percent Range 0.97
Packaging Glass bottle
Quantity 1 kg
Beilstein 02, 606
Merck Index 15, 8808
Solubility Information Solubility in water: 270g/L water (25°C). Other solubilities: insoluble in alcohol
Formula Weight 270.13
Percent Purity 0.97
Insoluble Matter 0.005% max.
Physical Form Liquid
Chemical Name or Material Succinic acid, disodium salt hexahydrate, 0.97
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.