Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-Trichlorobenzene, for GPC, GC and spectrophotometry, >98.0%, Solstice

Catalog No. 6000071 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-000-71 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-000-71 Supplier Solstice Supplier No. 3484
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 120-82-1 , 108-70-3 , 87-61-6
Molecular Formula C6H3Cl3
Molecular Weight (g/mol) 181.44
InChI Key PBKONEOXTCPAFI-UHFFFAOYSA-N
Synonym benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene
PubChem CID 13
ChEBI CHEBI:28222
IUPAC Name 1,2,4-trichlorobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)Cl

Specifications

Absorbance 0.5 AU max. at 310 nm, 0.05 AU max. at 350 nm, 0.01 AU max. at 375 nm, 0.005 AU max. at 400 nm
Boiling Point 213.5°C
CAS Max % 98.0000%, <2.0000%
UN Number UN2321
Beilstein 956819
Density 1.454 g/cm3
Formula Weight 181.46
Percent Purity >98%
Grade Spectrophotometry
Packaging Glass Bottle
Quantity 4 x 4 L
Flash Point 105°C
Chemical Name or Material 1,2,4-Trichlorobenzene, B&J Brand™, for GPC, GC and spectrophotometry, >98.0%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Trichlorobenzenes, liquid
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.