Alfa Aesar™ 1,2,4-Trimethylbenzene, 98%

Manufacturer: Alfa Aesar™ 1241136

Quantity 500g

 View more versions of this product

Catalog No. AA1241136

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 95-63-6
    Formula Weight 120.2
    MDL Number MFCD00008527
    UN Number UN3295
    Beilstein 1903005
    Merck Index 14,7915
    Synonym Pseudocumene
    Chemical Name or Material 1,2,4-Trimethylbenzene

    Specifications

    Formula Weight 120.2
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Merck Index 14,7915
    Solubility Information Soluble in alcohol, benzene, ether
    Health Hazard 1 Warning
    Density 0.882g/mL
    Odor Aromatic
    Melting Point −44°C
    Refractive Index 1.5040
    Physical Form Liquid
    MDL Number MFCD00008527
    UN Number UN3295
    Beilstein 1903005
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
    RTECS Number DC3325000
    Boiling Point 169° to 171°C
    TSCA TSCA
    CAS 95-63-6
    Health Hazard 2 GHS H Statement
    H226-H332-H315-H319-H335
    Flammable liquid and vapor.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Flash Point 48° (118°F)
    Quantity 500g
    Synonym Pseudocumene
    Chemical Name or Material 1,2,4-Trimethylbenzene
    Assay 98%

    GHS H Statement
    H226-H332-H315-H319-H335
    Flammable liquid and vapor.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit