Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,5,6-Diepoxycyclooctane, 96%, Thermo Scientific™

Catalog No. AC170900100
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 27035-39-8
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00005076
InChI Key USGYMDAUQBQWFU-UHFFFAOYSA-N
Synonym 1r,4r,6r,9s-5,10-dioxatricyclo 7.1.0.0?,? decane
PubChem CID 91447
SMILES C1CC2C(O2)CCC3C1O3

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 90.0°C (5.0 mmHg)
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4950 to 1.4980
Quantity 10 g
Formula Weight 140.18
Percent Purity 96%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 1,2,5,6-Diepoxycyclooctane, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Suspected of causing cancer.
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 248-180-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.