Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride, 98%, Alfa Aesar™

Click to view available options
Quantity:
5g
25g
100g
130198-05-9
C15H24ClNO2
285.812
MFCD06658143
NTKXIDDUCSFBBF-UHFFFAOYSA-N
1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol hydrochloride,1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol hydrochloride,1-2-amino-1-4-methoxyphenyl-ethyl-cyclohexanol hydrochloride,2-4-methoxyphenyl-2-1-hydroxycyclohexyl ethylamine hydrochloride,1-4-methoxyphenyl-2-aminoethyl cyclohexanol hydrochloride,n,n-didesmethylvenlafaxine hydrochloride,1-2-amino-1-4-methoxyphenyl ethyl cyclohexan-1-ol hydrochloride,1-2-amino-1-4-methyoxyphenyl ethyl cyclohexanol.hcl,1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol.hcl,1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol hcl
9795856
1-[2-amino-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol;hydrochloride
COC1=CC=C(C=C1)C(CN)C2(CCCCC2)O.Cl
130198-05-9
C15H24ClNO2
285.812
MFCD06658143
NTKXIDDUCSFBBF-UHFFFAOYSA-N
1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol hydrochloride,1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol hydrochloride,1-2-amino-1-4-methoxyphenyl-ethyl-cyclohexanol hydrochloride,2-4-methoxyphenyl-2-1-hydroxycyclohexyl ethylamine hydrochloride,1-4-methoxyphenyl-2-aminoethyl cyclohexanol hydrochloride,n,n-didesmethylvenlafaxine hydrochloride,1-2-amino-1-4-methoxyphenyl ethyl cyclohexan-1-ol hydrochloride,1-2-amino-1-4-methyoxyphenyl ethyl cyclohexanol.hcl,1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol.hcl,1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol hcl
9795856
1-[2-amino-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol;hydrochloride
COC1=CC=C(C=C1)C(CN)C2(CCCCC2)O.Cl
Quantity 5g
Color White
Melting Point 174°C to 178°C (literature)
Beilstein 4215444
Formula Weight 285.81
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning