Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium carbonate, 99+%, ACS reagent

Catalog No. AC413260025
Click to view available options
Quantity:
100 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle

CAS: 554-13-2 | CLi2O3 | 73.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
IUPAC Name dilithium(1+) carbonate
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Melting Point 720.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 99+%
Linear Formula Li2CO3
Fieser 01,606; 03,183; 04,192; 05,396
Merck Index 15,5583
Solubility Information Solubility in water: 13g/L (20°C). Other solubilities: soluble in diluted acids, practically insoluble in alcohol
Formula Weight 73.88
Percent Purity ≥99%
Grade ACS Reagent
Packaging Plastic Bottle
Density 2.11g/mL
Chemical Name or Material Lithium carbonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 209-062-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.