Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1,2-Benzenedimethanol, 97%
Click to view available options
Quantity:
1 g
5 g
25 g
Description
1,2-Benzenedimethanol is a raw material used in chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2-Benzenedimethanol is a raw material used in chemical synthesis.
Solubility
Soluble in water, ether, ethanol, benzene.
Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
1,2-Benzenedimethanol is a raw material used in chemical synthesis.
Solubility
Soluble in water, ether, ethanol, benzene.
Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 59°C to 64°C |
| Quantity | 25 g |
| Beilstein | 508775 |
| Solubility Information | Soluble in water,ether,ethanol,benzene. |
| Formula Weight | 138.17 |
| Percent Purity | 97% |
| Chemical Name or Material | 1,2-Benzenedimethanol |
Safety and Handling
| EINECSNumber | 210-293-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction