Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Benzenedimethanol, 97%

Catalog No. p-7026787
Click to view available options
Quantity:
1 g
5 g
25 g

Catalog No. AAB2300803

Supplier: Thermo Scientific Chemicals B2300803

Only null left
Add to Cart

CAS: 612-14-6 | C8H10O2 | 138.17 g/mol

1,2-Benzenedimethanol is a raw material used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Benzenedimethanol is a raw material used in chemical synthesis.

Solubility
Soluble in water, ether, ethanol, benzene.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-14-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00004626
InChI Key XMUZQOKACOLCSS-UHFFFAOYSA-N
Synonym 1,2-benzenedimethanol, 1,2-phenylenedimethanol, phthalyl alcohol, 1,2-dihydroxymethylbenzene, o-xylene-alpha,alpha'-diol, 2-hydroxymethyl phenyl methanol, benzenedimethanol, o-xylylene glycol, 1,2-di hydroxymethyl benzene, 2-hydroxymethylbenzenemethanol
PubChem CID 69153
IUPAC Name [2-(hydroxymethyl)phenyl]methanol
SMILES OCC1=CC=CC=C1CO

Specifications

Melting Point 59°C to 64°C
Quantity 1 g
Beilstein 508775
Solubility Information Soluble in water,ether,ethanol,benzene.
Formula Weight 138.17
Percent Purity 97%
Chemical Name or Material 1,2-Benzenedimethanol
EINECSNumber 210-293-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.