Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Benzenedithiol, 96%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
17534-15-5
C6H6S2
142.2
MFCD00004835
JRNVQLOKVMWBFR-UHFFFAOYSA-N
1,2-benzenedithiol,benzenedithiol,1,2-dimercaptobenzene,dithiocatechol,dimercaptobenzene,o-benzenedithiol,acmc-209e9y,1,2-benzenedithiol 9ci,benzene-1,2-dithiol
69370
benzene-1,2-dithiol
C1=CC=C(C(=C1)S)S
17534-15-5
C6H6S2
142.2
MFCD00004835
JRNVQLOKVMWBFR-UHFFFAOYSA-N
1,2-benzenedithiol,benzenedithiol,1,2-dimercaptobenzene,dithiocatechol,dimercaptobenzene,o-benzenedithiol,acmc-209e9y,1,2-benzenedithiol 9ci,benzene-1,2-dithiol
69370
benzene-1,2-dithiol
C1=CC=C(C(=C1)S)S
Melting Point 27.0°C to 28.0°C
Infrared Spectrum Authentic
Color Yellow
Packaging Glass bottle
Refractive Index 1.6565 to 1.6585
Quantity 5g
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Solubility Information Solubility in water: insoluble
Formula Weight 142.2
Physical Form Liquid After Melting
Percent Purity 96%
Chemical Name or Material 1, 2-Benzenedithiol
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a

GHS Signal Word: Warning

EINECSNumber : 241-530-4