Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1,2-Benzofluorene 95.0+%, TCI America™

Click to view available options
Quantity:
200 mg
1 g
Specifications
Specifications
Melting Point | 182°C |
Boiling Point | 407°C |
Quantity | 1 g |
UN Number | 3077 |
Formula Weight | 216.28 |
Percent Purity | ≥95.0% (GC) |
Physical Form | Crystalline Powder |
Chemical Name or Material | 1,2-Benzofluorene |
Safety and Handling
RTECSNumber | DF6382000 |
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction