Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(chlorodimethylsilyl)ethane 95.0+%, TCI America™
SDP

Catalog No. B168825G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 13528-93-3
Molecular Formula C6H16Cl2Si2
Molecular Weight (g/mol) 215.26
MDL Number MFCD00000505
InChI Key VGQOKOYKFDUPPJ-UHFFFAOYSA-N
Synonym 1,2-bis chlorodimethylsilyl ethane, ethylenebis chlorodimethylsilane, silane, 1,2-ethanediylbis chlorodimethyl, unii-b4ocj0e6j1, 1,1,4,4-tetramethyl-1,4-dichlorodisilethylene, chloro 2-chlorodimethylsilyl ethyl dimethylsilane, b4ocj0e6j1, 2,5-dichloro-2,5-dimethyl-2,5-disilahexane, 1,1,4,4-tetramethyl-1,4-dichloro-disylethylene
PubChem CID 83552
IUPAC Name chloro[2-(chlorodimethylsilyl)ethyl]dimethylsilane
SMILES C[Si](C)(Cl)CC[Si](C)(C)Cl

Specifications

Color White
Boiling Point 198°C
Quantity 25 g
UN Number 2987
Formula Weight 215.26
Percent Purity ≥95.0% (GC)
Physical Form Crystals
Chemical Name or Material 1,2-Bis(chlorodimethylsilyl)ethane [Protecting Reagent for Primary Amines]
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.