Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(chlorodimethylsilyl)ethane, tech. 90%

Catalog No. AAL0257022
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0257022 100 g
AAL0257014 25 g
AAL0257036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0257022 Supplier Thermo Scientific Chemicals Supplier No. L0257022
Add to Cart

CAS: 13528-93-3 | C6H16Cl2Si2 | 215.26 g/mol

1,2-Bis(chlorodimethylsilyl)ethane is used as Protecting Reagent for Primary Amines. It is also used in Medicine field, electronics industry and polymer material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis(chlorodimethylsilyl)ethane is used as Protecting Reagent for Primary Amines. It is also used in Medicine field, electronics industry and polymer material.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Incompatible with Strong oxidizing agents, Strong bases and water. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13528-93-3
Molecular Formula C6H16Cl2Si2
Molecular Weight (g/mol) 215.26
MDL Number MFCD00000505
InChI Key VGQOKOYKFDUPPJ-UHFFFAOYSA-N
Synonym 1,2-bis chlorodimethylsilyl ethane, ethylenebis chlorodimethylsilane, silane, 1,2-ethanediylbis chlorodimethyl, unii-b4ocj0e6j1, 1,1,4,4-tetramethyl-1,4-dichlorodisilethylene, chloro 2-chlorodimethylsilyl ethyl dimethylsilane, b4ocj0e6j1, 2,5-dichloro-2,5-dimethyl-2,5-disilahexane, 1,1,4,4-tetramethyl-1,4-dichloro-disylethylene
PubChem CID 83552
SMILES C[Si](C)(Cl)CC[Si](C)(C)Cl

Specifications

Boiling Point 198°C to 200°C
Melting Point 34°C to 40°C
Quantity 100 g
UN Number UN3261
Beilstein 1920141
Flash Point 65°C (149°F)
Solubility Information Reacts with water.
IUPAC Name chloro-[2-[chloro(dimethyl)silyl]ethyl]-dimethylsilane
Formula Weight 215.27
Percent Purity 90%
Grade Technical
Sensitivity Moisture sensitive
Chemical Name or Material 1,2-Bis(chlorodimethylsilyl)ethane
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 236-871-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.