Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(dimethylsilyl)benzene, 98%

Catalog No. p-7059004
Click to view available options
Quantity:
2 g
10 g

CAS: 17985-72-7 | C10H16Si2 | 192.408 g/mol

1,2-Bis(dimethylsilyl)benzene is a protecting group-reagent for amines and amino acids forming benzostabase derivatives. It is an important raw material and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17985-72-7
Molecular Formula C10H16Si2
Molecular Weight (g/mol) 192.408
MDL Number MFCD00142462
InChI Key MUUXBTFQEXVEEI-UHFFFAOYSA-N
Synonym 1,2-bis dimethylsilyl benzene, 1,2-phenylenebis dimethylsilane, 2-dimethyl, acmc-209efy, bis dimethylsilyl benzene, amtsi084, o-phenylenebis dimethylsilane, 2-dimethylsilylphenyl-dimethylsilane, 5,6-bis dimethylsilanediyl-1,3-cyclohexadiene
PubChem CID 6364986
IUPAC Name [2-(dimethyl-$l^{3}-silanyl)phenyl]-dimethylsilicon
SMILES C[Si](C)C1=CC=CC=C1[Si](C)C

Specifications

Density 0.898 g/mL
Boiling Point 101°C (13 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.514
Quantity 2 g
Solubility Information Insoluble in water.
Formula Weight 194.43
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,2-Bis (dimethylsilyl)benzene
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.