Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(phenylthio)ethylene (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. B294025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B294025G 25 g
B29405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B294025G Supplier TCI America Supplier No. B294025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23528-44-1
Molecular Formula C14H12S2
Molecular Weight (g/mol) 244.37
InChI Key UHGSEDVQATYWGI-VAWYXSNFSA-N
Synonym 1,2-Bis(phenylmercapto)ethylene
PubChem CID 5372209
IUPAC Name [(E)-2-phenylsulfanylethenyl]sulfanylbenzene
SMILES C1=CC=C(C=C1)SC=CSC2=CC=CC=C2

Specifications

Melting Point 48°C
Boiling Point 166°C
Quantity 25 g
Formula Weight 244.37
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 1,2-Bis(phenylthio)ethylene (cis- and trans- mixture)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.