Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(phosphino)ethane, 99%

Catalog No. AA39279MD
Click to view available options
Quantity:
250 mg

CAS: 5518-62-7 | C2H8P2 | 94.034 g/mol

It is used in chemical research organic Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5518-62-7
Molecular Formula C2H8P2
Molecular Weight (g/mol) 94.034
MDL Number MFCD00049195
InChI Key FWFZRPMNAAFGBA-UHFFFAOYSA-N
Synonym 1,2-bis phosphino ethane, 2-phosphanylethyl phosphane, ethylenediphosphine, ethylenebisphosphine, 1,2-bis-phosphino ethane bpe, phosphine,1,2-ethanediylbis-9ci
PubChem CID 4361793
IUPAC Name 2-phosphanylethylphosphane
SMILES C(CP)P

Specifications

Color Colorless
Boiling Point 109°C to 110°C
Odor Pungent
Linear Formula H2PCH2CH2PH2
Quantity 250 mg
UN Number UN2845
Sensitivity Air Sensitive
Solubility Information It is soluble in organic solvents. It is insoluble in water.
Formula Weight 94.03
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,2-Bis(phosphino)ethane
Show More Show Less
Hazard Category H250-H252-H314-H335
Hazard Statement GHS H Statement
H250-H252-H314-H318
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P222-P235+P410-P260-P264b-P271-P280-P301+P330+P331-P302+P334-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.