Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(trimethoxysilyl)ethane, 96%

Catalog No. p-7053542
Click to view available options
Quantity:
5 mL
25 mL

CAS: 18406-41-2 | C8H22O6Si2 | 270.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18406-41-2
Molecular Formula C8H22O6Si2
Molecular Weight (g/mol) 270.43
MDL Number MFCD00053672
InChI Key JCGDCINCKDQXDX-UHFFFAOYSA-N
Synonym 1,2-bis trimethoxysilyl ethane, 3,3,6,6-tetramethoxy-2,7-dioxa-3,6-disilaoctane, hexamethoxydisilylethane, 1,2-ethylenebis trimethoxysilane, dow corning x1-6145a, 2,7-dioxa-3,6-disilaoctane, 3,3,6,6-tetramethoxy, trimethoxy 2-trimethoxysilylethyl silane, 1,4-disilabutane, 1,1,1,4,4,4-hexamethoxy, acmc-209ele, ethylenebis trimethoxysilane
PubChem CID 87627
SMILES CO[Si](CC[Si](OC)(OC)OC)(OC)OC

Specifications

Color Colorless
Density 1.073 g/mL
Boiling Point 103°C to 104°C
Flash Point 109°C (228°F)
Refractive Index 1.409
Linear Formula [-CH2Si(OCH3)3]2
Quantity 5 mL
UN Number UN3382
IUPAC Name trimethoxy(2-trimethoxysilylethyl)silane
Formula Weight 270.43
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1,2-Bis(trimethoxysilyl)ethane
Show More Show Less
Hazard Category H330
Hazard Statement GHS H Statement
H330
Fatal if inhaled.
Precautionary Statement P260-P271-P284-P304+P340-P310-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, N.O.S.
EINECSNumber 242-285-6
RTECSNumber JG7873000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.