Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Bromoethoxy)-4-chlorobenzene, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2033-76-3
C8H8BrClO
235.51
MFCD00000617
YYFLBDSMQRWARK-UHFFFAOYSA-N
1-2-bromoethoxy-4-chlorobenzene,benzene, 1-2-bromoethoxy-4-chloro,2-bromoethyl 4-chlorophenyl ether,4-chlorophenyl 2-bromoethyl ether,2-4-chlorophenoxy ethyl bromide,benzene,1-2-bromoethoxy-4-chloro,2-bromo-1-4-chlorophenoxy ethane,pubchem17076,acmc-1cjap,p-chlorophenoxyethyl bromide
74854
1-(2-bromoethoxy)-4-chlorobenzene
C1=CC(=CC=C1OCCBr)Cl
2033-76-3
C8H8BrClO
235.51
MFCD00000617
YYFLBDSMQRWARK-UHFFFAOYSA-N
1-2-bromoethoxy-4-chlorobenzene,benzene, 1-2-bromoethoxy-4-chloro,2-bromoethyl 4-chlorophenyl ether,4-chlorophenyl 2-bromoethyl ether,2-4-chlorophenoxy ethyl bromide,benzene,1-2-bromoethoxy-4-chloro,2-bromo-1-4-chlorophenoxy ethane,pubchem17076,acmc-1cjap,p-chlorophenoxyethyl bromide
74854
1-(2-bromoethoxy)-4-chlorobenzene
C1=CC(=CC=C1OCCBr)Cl
Melting Point 38°C to 39°C
Flash Point 150°C
Boiling Point 150°C to 152°C (18.0mmHg)
Quantity 25g
Color Undesignated
Refractive Index 1.5570 to 1.5590 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula ClC6H4O(CH2)2Br
Formula Weight 235.51
Physical Form Liquid or Low Melting Solid
Percent Purity 99%
Chemical Name or Material 1-(2-Bromoethoxy)-4-chlorobenzene
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-994-9