Learn More
1,2-Cyclohexanedione dioxime, 97%
Description
1,2-Cyclohexanedione dioxime acts as a chelating ligand forming complexes with many transition metal ions like Co, Fe and Ni. It is used in photometric detection of metal ions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,2-Cyclohexanedione dioxime acts as a chelating ligand forming complexes with many transition metal ions like Co, Fe and Ni. It is used in photometric detection of metal ions.
Solubility
Soluble in water.
Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.

Specifications
Specifications
Melting Point | 181°C to 188°C (decomposition) |
Quantity | 25 g |
Beilstein | 1909047 |
Merck Index | 14,6560 |
Solubility Information | Soluble in water. |
Formula Weight | 142.16 |
Percent Purity | 97% |
Chemical Name or Material | 1,2-Cyclohexanedione dioxime |
Safety and Handling
EINECSNumber | 207-769-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.