Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Cyclohexanedione 97.0+%, TCI America™
SDP

Catalog No. C072825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C072825G 25 g
C07285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C072825G Supplier TCI America Supplier No. C072825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 765-87-7
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00001648
InChI Key OILAIQUEIWYQPH-UHFFFAOYSA-N
Synonym 1,2-cyclohexanedione, 1,2-dioxocyclohexane, cyclohexanedione, 1,2-cyclohexadione, cyclohexan-1,2-dione, ccris 6296, unii-75c1ovw0fj, 75c1ovw0fj, 1,2-cyclohexanedione,ketone form, dihydrocatechol
PubChem CID 13006
ChEBI CHEBI:41674
IUPAC Name cyclohexane-1,2-dione
SMILES C1CCC(=O)C(=O)C1

Specifications

Color Yellow
Boiling Point 194°C
Quantity 25 g
Formula Weight 112.13
Percent Purity ≥97.0% (GC)
Physical Form Crystals
Chemical Name or Material 1,2-Cyclohexanedione
RTECSNumber GV0340000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.