Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Decanediol 99.0+%, TCI America™
SDP

Catalog No. D2720500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2720500G 500 g
D272025G 25 g
D2720100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D2720500G Supplier TCI America Supplier No. D2720500G

Chemical Identifiers

CAS 1119-86-4
Molecular Formula C10H22O2
Molecular Weight (g/mol) 174.284
MDL Number MFCD00010739
InChI Key YSRSBDQINUMTIF-UHFFFAOYSA-N
Synonym 1,2-decanediol, decylene glycol, decane 1,2-diol, 1,2-dihydroxydecane, acmc-20apfz, acmc-20apjb, pubchem12680, acmc-1bny3
PubChem CID 79141
IUPAC Name decane-1,2-diol
SMILES CCCCCCCCC(CO)O

Specifications

Melting Point 49°C
Color White
Boiling Point 168°C
Quantity 500 g
Formula Weight 174.28
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Decanediol
EINECSNumber (2)-0240
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.