Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Diaminocyclohexanetetraacetic acid monohydrate, Solstice

Catalog No. 6002542 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-025-42 100 g
60-025-43 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-025-42 Supplier Solstice Supplier No. 34588100G
Add to Cart
Add to Cart

for complexometry,≥98.5%

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145819-99-4
Molecular Formula C14H24N2O9
Molecular Weight (g/mol) 364.351
MDL Number MFCD00150952
InChI Key VASZYFIKPKYGNC-UHFFFAOYSA-N
Synonym cdta hydrate, glycine, n,n'-1,2-cyclohexanediylbis n-carboxymethyl-, monohydrate, trans-1,2-cyclohexanediamine-n,n,n',n'-tetraacetic acid monohydrate, acmc-20n4nc, 1,2-diaminocyclohexanetetraacetic acid monohydrate, trans-1,2-cyclohexanediamine-n,n,n',n-tetraacetic acid, 2,2',2,2'-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, 2,2',2,2'-cyclohexane-1,2-diyldinitrilo tetraacetic acid-water 1/1, 2-2-bis carboxymethyl amino cyclohexyl-carboxymethyl amino acetic acid hydrate, 1,2-diaminocyclohexanetetraacetic acid monohydrate for complexometry, inverted exclamation marky
PubChem CID 18674933
IUPAC Name 2-[[2-[bis(carboxymethyl)amino]cyclohexyl]-(carboxymethyl)amino]acetic acid;hydrate
SMILES C1CCC(C(C1)N(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O

Specifications

Quantity 100 g
UN Number NONH for all modes of transport
Beilstein 6884761
Formula Weight 364.35g/mol
Percent Purity ≥98.5%
Chemical Name or Material 1,2-Diaminocyclohexanetetraacetic acid monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.