Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Diaminopropane, 99%

Catalog No. p-3736111
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle

CAS: 78-90-0 | C3H10N2 | 74.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-90-0
Molecular Formula C3H10N2
Molecular Weight (g/mol) 74.13
MDL Number MFCD00008089
InChI Key AOHJOMMDDJHIJH-UHFFFAOYNA-N
Synonym 1,2-diaminopropane, propylenediamine, 1,2-propanediamine, propylene diamine, 2,3-diaminopropane, 1,2-propylenediamine, 1,2 diaminopropane, ccris 4863, 1-methylethylenediamine, 2-aminopropylamine
PubChem CID 6567
ChEBI CHEBI:30630
IUPAC Name propane-1,2-diamine
SMILES CC(N)CN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -37.0°C
Density 0.8700g/mL
Boiling Point 119.0°C to 120.0°C
Flash Point 33°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4450 to 1.4470
Linear Formula CH3CH(NH2)CH2NH2
Quantity 250 mL
Beilstein 04,257
Specific Gravity 0.87
Merck Index 15,7965
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in ether, soluble in acetone, benzene, chloroform and, ethanol
Formula Weight 74.13
Percent Purity 99%
Chemical Name or Material 1, 2-Diaminopropane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
Flammable liquid and vapor.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cau
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
EINECSNumber 201-155-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.