Alfa Aesar™ 1,2-Dianilinoethane, 98+%

Manufacturer: Alfa Aesar™ B2309814

Quantity 25g

 View more versions of this product

Catalog No. AAB2309814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 150-61-8
    Molecular Formula C6H5NHCH2CH2NHC6H5
    Formula Weight 212.3
    MDL Number MFCD00003019
    Beilstein 646740
    Merck Index 14,2990
    Synonym N,N'-Diphenylethylenediamine; Wanzlick's Reagent
    Chemical Name or Material 1,2-Dianilinoethane

    Specifications

    Formula Weight 212.3
    Melting Point 64° to 68°C
    Quantity 25g
    MDL Number MFCD00003019
    Merck Index 14,2990
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    RTECS Number KV4800000
    TSCA TSCA
    Boiling Point 228°C (12mmHg)
    Molecular Formula C6H5NHCH2CH2NHC6H5
    CAS 150-61-8
    Beilstein 646740
    Synonym N,N'-Diphenylethylenediamine; Wanzlick's Reagent
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 1,2-Dianilinoethane
    Assay ≥98%

    Description

    Applications
    1,2-Dianilinoethane can forms crystalline imidazolidines with aldehydes in the presence of AcOH. N-donating ligand. Can be used as a trapping agent for aldehydes.

    Solubility
    Sparingly soluble in water 0.072 g/L @ 25°C.

    Notes
    Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

    GHS H Statement
    H301-H311-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit