Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dibromo-4,5-difluorobenzene 97.0+%, TCI America™
SDP

Catalog No. D191325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D191325G 25 g
D19135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D191325G Supplier TCI America Supplier No. D191325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64695-78-9
Molecular Formula C6H2Br2F2
Molecular Weight (g/mol) 271.887
MDL Number MFCD00009890
InChI Key JTEZQWOKRHOKDG-UHFFFAOYSA-N
Synonym benzene, 1,2-dibromo-4,5-difluoro, 1,2-dibromo-4,5-difluoro-benzene, 1,2-difluoro-4,5-dibromobenzene, zlchem 536, pubchem3440, acmc-1biw8, 1,2-dibromo-4,5-difluorobenzene, benzene, 1,2-dibromo-4,5-difluoro-;1,2-dibromo-4,5-difl
PubChem CID 94949
IUPAC Name 1,2-dibromo-4,5-difluorobenzene
SMILES C1=C(C(=CC(=C1Br)Br)F)F

Specifications

Color White-Yellow
Quantity 25 g
UN Number 1325
Formula Weight 271.89
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,2-Dibromo-4,5-difluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.