Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichloroethane, 99.0% min., ACS, MilliporeSigma™

Catalog No. MDX08002 Shop All MilliporeSigma Products
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy

Catalog No. MDX08002

Supplier: MilliporeSigma DX08002

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 107-06-2
Molecular Formula C2H4Cl2
Molecular Weight (g/mol) 98.95
MDL Number MFCD00000963
InChI Key WSLDOOZREJYCGB-UHFFFAOYSA-N
Synonym ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
PubChem CID 11
ChEBI CHEBI:27789
IUPAC Name 1,2-dichloroethane
SMILES ClCCCl

Specifications

Color Colorless
Quantity 500 mL
Physical Form Liquid
Assay Percent Range 99.0% min.
Formula Weight 98.96
Chemical Name or Material 1, 2-Dichloroethane, For ACS analysis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.