Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichloroethane, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018892
Click to view available options
Quantity:
1 mL
5 mL
Featured Industries: Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 775°F
CAS 107-06-2
Vapor Pressure 87 mmHg (25°C)
Vapor Density 3.4 (20°C, vs. air)
Density 1.256 g/mL (at 25°C)
Flash Point 13°C - Closed Cup
Linear Formula ClCH2 CH2 Cl
MDL Number MFCD00000963
Melting Point -35°C
Boiling Point 83°C
Synonym Ethylene chloride; Ethylene dichloride
Type Analytical Standard
Quantity 5 mL
Formula Weight 98.96
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.