Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichloroethane, ACS, 99+%

Catalog No. p-3256791
Click to view available options
Quantity:
500 mL
1 L
4 x 1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Maximum level of impurities: Color (APHA) 10, Evaporation residue 0.002%, Titratable acid 0.0003meq/g, H2O 0.03% | CAS: 107-06-2 | C2H4Cl2 | 98.95 g/mol

Used to make vinyl chloride, and chlorinated solvents(trichloroethane, trichloroethylene, perchloroethylene, and vinylidene), remove grease, resins, glue and dirt. Used as a solvent in the manufacturing of polystyrene and SBR latex. It is added to leaded petrol as an anti-knock compound. It is also used in polyvinyl chloride (PVC) pipes, packaging materials, furniture, auto mobile parts, wall coverings and housewares. It is also used generally as an intermediate for other organic chemical compounds and as a solvent. Used as fumigant residues for wheat, flour, bran, middlings and bread. Used as solvent for the isomer distribution in the acetylation of benzene and toluene under the influence of aluminum chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-06-2
Molecular Formula C2H4Cl2
Molecular Weight (g/mol) 98.95
MDL Number MFCD00000963
InChI Key WSLDOOZREJYCGB-UHFFFAOYSA-N
Synonym ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
PubChem CID 11
ChEBI CHEBI:27789
IUPAC Name 1,2-dichloroethane
SMILES ClCCCl

Specifications

Melting Point °C to 35°C
Color Colorless
Appearance Colorless liquid
Density 1.256 g/cm3 at 20°C
Boiling Point 83°C
Quantity 500 mL
Flash Point 15°C (59°F)
Ignition Point 440°C (824°F)
Physical Form Liquid
Refractive Index 1.4448
Assay Percent Range >99%
Beilstein 605264
Solubility Information Miscible in methanol,diethyl ether,n-octanol,acetone. Very slightly miscible in water.
Formula Weight 98.96
Grade ACS
Concentration or Composition (by Analyte or Components) 1, 2-Dichloroethane
Chemical Name or Material 1, 2-Dichloroethane
Show More Show Less
Hazard Category H225-H302-H315-H319-H331-H335-H336-H350-H373
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P311-P330-P332+P313-P363-P370+P378q-P5
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 203-458-1
RTECSNumber KI0525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.