Learn More
1,2-Dichloroethane, Spectrophotometric Grade, 99+%
Description
1,2-dichloroethane is used in the production of vinyl chloride which is used to make a variety of plastic and vinyl products including polyvinyl chloride (PVC) pipes, furniture and automobile upholstery, wall coverings, house wares, and automobile parts.1,2-Dichloroethane is also used generally as an intermediate for other organic chemical compounds and as a solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -35°C |
| Density | 1.256 g/mL |
| Boiling Point | 83°C |
| Quantity | 4 L |
| Flash Point | 15°C (59°F) |
| Physical Form | Liquid |
| Refractive Index | 1.4448 |
| UN Number | UN1184 |
| Beilstein | 605264 |
| Merck Index | 14,3797 |
| Show More |
Safety and Handling
| Hazard Category | H225-H302-H315-H319-H331-H335-H336-H350-H373 |
| Hazard Statement | GHS H Statement H225-H350-H302-H315-H319-H335 Highly flammable liquid and vapor. May cause cancer. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P311-P330-P332+P313-P363-P370+P378q-P5 |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYLENE DICHLORIDE |
| EINECSNumber | 203-458-1 |
| RTECSNumber | KI0525000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.