Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,2-Dichloropropane

Catalog No. 501859799
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-185-9799 250 mL
50-185-9798 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-185-9799 Supplier Sigma Aldrich Supplier No. 82270250ML
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,2-Dichloropropane
TRUSTED_SUSTAINABILITY

Specifications

CAS 78-87-5
Melting Point -100°C (lit.)
Density 1.156 g/mL (at 25°C (literature))
Boiling Point 95°C to 96°C (lit.)
Molecular Formula C3H6Cl2
Refractive Index n20/D 1.439 (literature); n20/D 1.439
Linear Formula CH3CHClCH2Cl
MDL Number MFCD00000868
Quantity 250 mL
Synonym Propylene dichloride
Molecular Weight (g/mol) 112.99
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.