Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichlorotetramethyldisilane 95.0+%, TCI America™
SDP

Catalog No. D214425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D214425G 25 g
D21445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D214425G Supplier TCI America Supplier No. D214425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4342-61-4
Molecular Formula C4H12Cl2Si2
Molecular Weight (g/mol) 187.21
MDL Number MFCD00078528
InChI Key SFAZXBAPWCPIER-UHFFFAOYSA-N
PubChem CID 78045
IUPAC Name 1,2-dichloro-1,1,2,2-tetramethyldisilane
SMILES C[Si](C)(Cl)[Si](C)(C)Cl

Specifications

Color Yellow
Boiling Point 148°C
Quantity 25 g
UN Number 2986
Formula Weight 187.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Dichlorotetramethyldisilane
EINECSNumber (2)-2040
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.