Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dimethyl-5-nitroimidazole, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
1kg
Packaging:
Glass bottle
551-92-8
C5H7N3O2
141.13
MFCD00047046
IBXPYPUJPLLOIN-UHFFFAOYSA-N
dimetridazole, 1,2-dimethyl-5-nitro-1h-imidazole, dimetridazol, emtrylvet, emtrymix, emtryl, unizole soluble, 1h-imidazole, 1,2-dimethyl-5-nitro, 5-nitro-1,2-dimethylimidazole, dimetridazolo
3090
1,2-dimethyl-5-nitroimidazole
CN1C(C)=NC=C1[N+]([O-])=O
551-92-8
C5H7N3O2
141.13
MFCD00047046
IBXPYPUJPLLOIN-UHFFFAOYSA-N
dimetridazole, 1,2-dimethyl-5-nitro-1h-imidazole, dimetridazol, emtrylvet, emtrymix, emtryl, unizole soluble, 1h-imidazole, 1,2-dimethyl-5-nitro, 5-nitro-1,2-dimethylimidazole, dimetridazolo
3090
1,2-dimethyl-5-nitroimidazole
CN1C(C)=NC=C1[N+]([O-])=O
Infrared Spectrum Authentic
Quantity 1kg
Color Cream to Yellow
Packaging Glass bottle
Melting Point 139.0°C to 140.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Merck Index 15,329
Formula Weight 141.13
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 1, 2-Dimethyl-5-nitroimidazole
Show More Show Less

GHS H Statement:
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-001-2