Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Epoxyhexane 96.0+%, TCI America™
SDP

Catalog No. E0445500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0445500ML 500 mL
E044525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0445500ML Supplier TCI America Supplier No. E0445500ML

Chemical Identifiers

CAS 1436-34-6
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00005154
InChI Key WHNBDXQTMPYBAT-UHFFFAOYSA-N
Synonym 1,2-epoxyhexane, butyloxirane, oxirane, butyl, 1-hexene oxide, 1-hexene epoxide, 1,2-hexene oxide, epoxy-n-hexane, hexylene oxide, hexane, 1,2-epoxy, hexene, monooxide
PubChem CID 15036
IUPAC Name 2-butyloxirane
SMILES CCCCC1CO1

Specifications

Color Yellow
Boiling Point 120°C
Quantity 500 mL
UN Number 1993
Formula Weight 100.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Epoxyhexane
EINECSNumber (2)-0220
RTECSNumber MO3630000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.