Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Epoxyhexane, 97%

Catalog No. AC335550050
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle

CAS: 1436-34-6 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1436-34-6
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00005154
InChI Key WHNBDXQTMPYBAT-UHFFFAOYSA-N
Synonym 1,2-epoxyhexane, butyloxirane, oxirane, butyl, 1-hexene oxide, 1-hexene epoxide, 1,2-hexene oxide, epoxy-n-hexane, hexylene oxide, hexane, 1,2-epoxy, hexene, monooxide
PubChem CID 15036
IUPAC Name 2-butyloxirane
SMILES CCCCC1CO1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.8300g/mL
Boiling Point 118°C to 120°C
Flash Point 15°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4040 to 1.407
Quantity 5 mL
Specific Gravity 0.83
Formula Weight 100.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 2-Epoxyhexane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 215-864-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.