Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Ethanedithiol, 95%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
Packaging:
Glass bottle
540-63-6
C2H6S2
94.19
MFCD00004892
VYMPLPIFKRHAAC-UHFFFAOYSA-N
1,2-ethanedithiol,1,2-dimercaptoethane,dithioglycol,ethylene mercaptan,ethylenedithiol,1,2-ethanethiol,ethylenedimercaptan,dithioethyleneglycol,ethylene dithioglycol,s-ethylene dimercaptan
10902
ethane-1,2-dithiol
C(CS)S
540-63-6
C2H6S2
94.19
MFCD00004892
VYMPLPIFKRHAAC-UHFFFAOYSA-N
1,2-ethanedithiol,1,2-dimercaptoethane,dithioglycol,ethylene mercaptan,ethylenedithiol,1,2-ethanethiol,ethylenedimercaptan,dithioethyleneglycol,ethylene dithioglycol,s-ethylene dimercaptan
10902
ethane-1,2-dithiol
C(CS)S
Packaging Glass bottle
Flash Point 50°C
Quantity 1L
Specific Gravity 1.123
Density 1.1230g/mL
Infrared Spectrum Authentic
Melting Point -41.0°C
Color Undesignated
Boiling Point 144.0°C to 146.0°C
Refractive Index 1.558
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula HSCH2CH2SH
Beilstein 01, 471
Fieser 01,356; 02,189; 05,290
Merck Index 15, 3780
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkalies, soluble in alcohol
Formula Weight 94.19
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 1, 2-Ethanedithiol
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Harmful in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger