Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Ethanedithiol, 95%

Catalog No. p-3200939
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle

CAS: 540-63-6 | C2H6S2 | 94.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-63-6
Molecular Formula C2H6S2
Molecular Weight (g/mol) 94.19
MDL Number MFCD00004892
InChI Key VYMPLPIFKRHAAC-UHFFFAOYSA-N
Synonym 1,2-ethanedithiol, 1,2-dimercaptoethane, dithioglycol, ethylene mercaptan, ethylenedithiol, 1,2-ethanethiol, ethylenedimercaptan, dithioethyleneglycol, ethylene dithioglycol, s-ethylene dimercaptan
PubChem CID 10902
IUPAC Name ethane-1,2-dithiol
SMILES C(CS)S

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -41.0°C
Density 1.1230g/mL
Boiling Point 144.0°C to 146.0°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.558
Linear Formula HSCH2CH2SH
Quantity 250 mL
Beilstein 01, 471
Fieser 01,356; 02,189; 05,290
Specific Gravity 1.123
Merck Index 15, 3780
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkalies, soluble in alcohol
Formula Weight 94.19
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1, 2-Ethanedithiol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove
EINECSNumber 208-752-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.