Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Furoyl)piperazine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. F070525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F070525G 25 g
F07055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F070525G Supplier TCI America Supplier No. F070525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60548-09-6
Molecular Formula C9H13ClN2O2
Molecular Weight (g/mol) 216.67
MDL Number MFCD00236012
InChI Key FMFUHCXDFVDINI-UHFFFAOYSA-N
Synonym 1-[(2-Furan)carbonyl]piperazine Hydrochloride
PubChem CID 12289910
IUPAC Name 1-(furan-2-carbonyl)piperazine hydrochloride
SMILES Cl.O=C(N1CCNCC1)C1=CC=CO1

Specifications

Melting Point 207°C
Quantity 25 g
Formula Weight 216.67
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Furoyl)piperazine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.