Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Hydroxyethyl)-4-methylpiperazine, 98%, Acros Organics

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass bottle
5464-12-0
C7H16N2O
144.22
MFCD03840691
QHTUMQYGZQYEOZ-UHFFFAOYSA-N
1-2-hydroxyethyl-4-methylpiperazine,2-4-methylpiperazin-1-yl ethanol,1-piperazineethanol, 4-methyl,2-4-methylpiperazin-1-yl ethan-1-ol,1-n-hydroxyethyl-4-methyl piperazine,2-4-methyl-piperazin-1-yl-ethanol,1-2-hydroxyethyl-4-methyl-piperazine,4-methylpiperazine-1-ethanol,2-4-methylpiperazinyl ethan-1-ol,2-4-methyl-1-piperazinyl ethanol
231184
2-(4-methylpiperazin-1-yl)ethanol
CN1CCN(CC1)CCO
5464-12-0
C7H16N2O
144.22
MFCD03840691
QHTUMQYGZQYEOZ-UHFFFAOYSA-N
1-2-hydroxyethyl-4-methylpiperazine,2-4-methylpiperazin-1-yl ethanol,1-piperazineethanol, 4-methyl,2-4-methylpiperazin-1-yl ethan-1-ol,1-n-hydroxyethyl-4-methyl piperazine,2-4-methyl-piperazin-1-yl-ethanol,1-2-hydroxyethyl-4-methyl-piperazine,4-methylpiperazine-1-ethanol,2-4-methylpiperazinyl ethan-1-ol,2-4-methyl-1-piperazinyl ethanol
231184
2-(4-methylpiperazin-1-yl)ethanol
CN1CCN(CC1)CCO
Boiling Point 54°C to 55°C (0.5 mbar)
Refractive Index 1.4850 to 1.487
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Color Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 144.22
Physical Form Low Melting Solid
Percent Purity ≥97.5%
Chemical Name or Material 1-(2-Hydroxyethyl)-4-methylpiperazine
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.