Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Hydroxyethyl)-4-methylpiperazine, 98%

Catalog No. AC384830050
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle

CAS: 5464-12-0 | C7H16N2O | 144.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5464-12-0
Molecular Formula C7H16N2O
Molecular Weight (g/mol) 144.22
MDL Number MFCD03840691
InChI Key QHTUMQYGZQYEOZ-UHFFFAOYSA-N
Synonym 1-2-hydroxyethyl-4-methylpiperazine, 2-4-methylpiperazin-1-yl ethanol, 1-piperazineethanol, 4-methyl, 2-4-methylpiperazin-1-yl ethan-1-ol, 1-n-hydroxyethyl-4-methyl piperazine, 2-4-methyl-piperazin-1-yl-ethanol, 1-2-hydroxyethyl-4-methyl-piperazine, 4-methylpiperazine-1-ethanol, 2-4-methylpiperazinyl ethan-1-ol, 2-4-methyl-1-piperazinyl ethanol
PubChem CID 231184
IUPAC Name 2-(4-methylpiperazin-1-yl)ethanol
SMILES CN1CCN(CC1)CCO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Boiling Point 54°C to 55°C (0.5 mbar)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4850 to 1.487
Quantity 5 g
Formula Weight 144.22
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material 1-(2-Hydroxyethyl)-4-methylpiperazine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.