Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[2-(morpholin-4-ylmethyl)phenyl]methanamine, 97%, Thermo Scientific™

Catalog No. CC48613CB
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC48613CB 250 mg Amber Glass Bottle
CC48613DA 1 g Amber Glass Bottle
CC48613EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC48613CB Supplier Thermo Scientific Chemicals Supplier No. CC48613CB
Add to Cart

Chemical Identifiers

CAS 91271-82-8
Molecular Formula C12H18N2O
Molecular Weight (g/mol) 206.289
InChI Key NMFAEZHWSZZJOA-UHFFFAOYSA-N
Synonym 2-morpholin-4-ylmethylbenzylamine, 1-2-morpholin-4-ylmethyl phenyl methanamine, 2-morpholin-4-ylmethyl-benzylamine, 1-2-morpholin-4-ylmethyl phenyl methylamine, 2-morpholinomethyl phenyl methanamine, 2-morpholin-4-ylmethyl phenyl methanamine, 2-morpholin-4-ylmethyl phenyl methylamine, 2-morpholin-4-ylmethylphenylamine, 2-morpholin-4-ylmethyl benzyl amine
PubChem CID 4962390
IUPAC Name [2-(morpholin-4-ylmethyl)phenyl]methanamine
SMILES C1COCCN1CC2=CC=CC=C2CN

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 206.29
Percent Purity 95%
Chemical Name or Material 1-[2-(Morpholin-4-ylmethyl)phenyl]methanamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.