Learn More
1,2-Naphthoquinone-4-sulfonic acid sodium salt, 97%
Description
Colorimetric determination of amino acids and amines1,2-Naphthoquinone-4-sulfonic acid sodium salt is used for the colorimetric determination of amino acids, amines and procaine hydrochloride in pharmaceutical preparations. It is also used in the characterization of phanerochaete chrysosporium. Further, it acts as an intermediate in the synthesis of carbohydrate based napthoquinones, which acts as anticancer agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Colorimetric determination of amino acids and amines1,2-Naphthoquinone-4-sulfonic acid sodium salt is used for the colorimetric determination of amino acids, amines and procaine hydrochloride in pharmaceutical preparations. It is also used in the characterization of phanerochaete chrysosporium. Further, it acts as an intermediate in the synthesis of carbohydrate based napthoquinones, which acts as anticancer agents.
Solubility
Soluble in water and acetone. Slightly soluble in alcohol and ethanol. Insoluble in chloroform.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∼289°C (decomposition) |
| Quantity | 100 g |
| Beilstein | 4169788 |
| Merck Index | 14,8646 |
| Solubility Information | Soluble in water and acetone; Slightly soluble in alcohol and ethanol; Insoluble in chloroform. |
| Formula Weight | 260.2 |
| Percent Purity | 97% |
| Chemical Name or Material | 1,2-Naphthoquinone-4-sulfonic acid sodium salt |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 208-308-9 |
| RTECSNumber | QK1490000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.