Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Naphthoquinone, 95%, Tech.

Catalog No. p-170757
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Packaging:
Glass bottle

CAS: 524-42-5 | C10H6O2 | 158.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 524-42-5
Molecular Formula C10H6O2
Molecular Weight (g/mol) 158.16
MDL Number MFCD00001698
InChI Key KETQAJRQOHHATG-UHFFFAOYSA-N
Synonym 1,2-naphthoquinone, 1,2-naphthalenedione, o-naphthoquinone, beta-naphthoquinone, 1,2-naphthaquinone, 1,2-naftochinon, .beta.-naphthoquinone, 1,2-naftochinon czech, ccris 1558, 1,2-dihydronaphthalene-1,2-dione
PubChem CID 10667
ChEBI CHEBI:34055
IUPAC Name naphthalene-1,2-dione
SMILES O=C1C=CC2=CC=CC=C2C1=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 136.0°C to 141.0°C
Color Colorless or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 07, 709
Fieser 01,713
Merck Index 15, 6479
Solubility Information Solubility in water: negligible. Other solubilities: soluble in 5% naoh,5% nahco3 and conc. h2so4,soluble in alcohol,benzene and ether
Formula Weight 158.16
Percent Purity 95%
Grade Technical
Physical Form Crystalline Mass or Waxy Solid
Chemical Name or Material 1, 2-Naphthoquinone, tech., 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
EINECSNumber 208-360-2
RTECSNumber QL7000000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.