Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Nitrophenyl)pyrrolidine, 97%, Thermo Scientific™

Catalog No. AAH6398403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6398403 1 g
AAH6398406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6398403 Supplier Thermo Scientific Chemicals Supplier No. H6398403

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40832-79-9
Molecular Formula C10H12N2O2
Molecular Weight (g/mol) 192.218
MDL Number MFCD01604393
InChI Key DHTSBEQPOXRZBB-UHFFFAOYSA-N
Synonym 1-2-nitrophenyl pyrrolidine, 2-nitro-1-pyrrolidinobenzene, 1-2-nitro-phenyl-pyrrolidine, 2-nitrophenyl pyrrolidine, o-pyrrolidin-1-ylnitrobenzene, acmc-1cujb, dhtsbeqpoxrzbb-uhfffaoysa
PubChem CID 12414144
IUPAC Name 1-(2-nitrophenyl)pyrrolidine
SMILES C1CCN(C1)C2=CC=CC=C2[N+](=O)[O-]

Specifications

Quantity 1 g
Formula Weight 192.21
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(2-Nitrophenyl)pyrrolidine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.