Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Octanediol, 98+%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
1117-86-8
C8H18O2
146.23
MFCD00010738
AEIJTFQOBWATKX-UHFFFAOYSA-N
1,2-octanediol,1,2-dihydroxyoctane,1,2-octylene glycol,capyryl glycol,racemic 1,2-octanediol,acmc-2099as,dsstox_cid_16646,dsstox_rid_79289
14231
CHEBI:34056
octane-1,2-diol
CCCCCCC(CO)O
1117-86-8
C8H18O2
146.23
MFCD00010738
AEIJTFQOBWATKX-UHFFFAOYSA-N
1,2-octanediol,1,2-dihydroxyoctane,1,2-octylene glycol,capyryl glycol,racemic 1,2-octanediol,acmc-2099as,dsstox_cid_16646,dsstox_rid_79289
14231
CHEBI:34056
octane-1,2-diol
CCCCCCC(CO)O
Melting Point 28°C to 31°C
Viscosity 35.5 mPa.s (40°C)
Boiling Point 267°C
Density 0.9140g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 0.914
Quantity 50g
Flash Point 138°C
Color Colorless to White
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (GC)
Linear Formula CH3(CH2)5CH(OH)CH2OH
Beilstein 01, III, 2217
Solubility Information Solubility in water: 3g/L (20°C).
Formula Weight 146.23
Physical Form Low Melting Solid
Percent Purity 98+%
Chemical Name or Material 1, 2-Octanediol, 98+ %
Show More Show Less

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 214-254-7

TSCA : TSCA